Imprimer
Connecting kits for CHROMABOND® Flash columns
Fournisseur: MACHEREY-NAGEL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
730798 732903
MANA730798EA 164 EUR
MANA730798 MANA732903
Connecting kits for CHROMABOND® Flash columns
Matériel de chromatographie Chromatographie, Pièces de rechange
CHROMABOND® Flash connecting kits allow to use CHROMABOND® Flash RS and BT cartridges as stand-alone system with any pump, detection, fraction collector combination.
Informations de livraison: CHROMABOND® Flash Starter Kit consists of ¹/₈" PTFE tubing, I.Ø×L (1,5 mm×3 m), 5×¹/₄"-28 PP nuts, 5×¹/₈" ETFE ferrules, 5×¹/₄"-28 nylon unions, 2×¹/₄"-28 PP Luer locks female, 1×¹/₄"-28 PP Luer locks male and 1×¹/₄"-28 PP Luer tip male.
CHROMABOND® Flash Stand Alone Kit consists of 1×¹/₄"-28 PP Luer lock female, 1×¹/₄"-28 PP Luer lock male, 2×¹/₈" ETFE ferrules, 2×¹/₄"-28 nylon unions and 2×¹/₄"-28 PP nuts.