Imprimer
VWR®, IOS Objectives for Inverted Microscopes
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
630-1764EA 566 EUR
630-1764 630-2164 630-2165 630-2166 630-2167 630-2168 630-2719
VWR®, IOS Objectives for Inverted Microscopes
Microscope Objectives
These objectives are designed to be used with inverted microscopes.
  • High resolution and contrast
  • Ensure field flatness up to F.N. 22, based on infinity-corrected optical system
  • Long Working Distance provides a wider working space between the lens surface and the object
Ideal for the observation of culture specimens, clinical examinations and cell testing.