Imprimer
Thermostatically controlled cabinets, for OxiTop®
Fournisseur: WTW
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
208450 208452 208464 208466 208468
705-1202EA 2430 EUR
705-1202 705-1203 WTWA208464 WTWA208466 WTWA208468
Thermostatically controlled cabinets, for OxiTop®
Incubateurs Incubateurs DBO
These thermostatic cabinets are available in three sizes and different variants. Suited for BOD, OECD 301F, soil respiration and anaerobic degradation.
  • Incubation conforming to standards
  • Temperature is adjustable in steps of 1 K between 10 and 40 °C
  • Temperature stability: ±0,5 K
  • Air circulation: 120 m³/h by means of tangential blower
  • Eco-friendly refrigerant R600a
TS 608/2i: A compact thermostatic cabinet for up two OxiTop® IS 12 stirring platforms. It is easy to integrate into the working environment and perfectly suited for standard applications. It has two internal plugs for connecting stirring platforms.
TS 608-G/2i: As the TS 608/2i but with a door made of insulation glass. Allows visual checking of the OxiTop® heads without opening the door.
TS 608-G/4i: As TS 608/4i but with a door made of insulation glass. Allows visual checking of the OxiTop® heads without opening the door.
TS 608/4i: Thermostatic cabinet for extensive sample processing. Capable of simultaneous measurement of up to four IS 12 stirring platforms. It has four internal plugs for connecting stirring platforms.
TS 1008i: A thermostatic cabinet for particular applications. With extra space for special vessels e.g. biogas or soil respiration measurement. It has four internal plugs for connecting stirring platforms.