Imprimer
Eye and face shower with bowl, wall mounted
Fournisseur: BROEN-LAB
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
17528009 17533009
BROE17528009EA 1190 EUR
BROE17528009 BROE17533009
Eye and face shower with bowl, wall mounted
Matériel de premiers secours Douches
Eye/Face shower with two heads in brass with chemical resistant BROEN-LAB Polycoat and stainless steel bowl.
  • Reliable and efficient showers for eyes and face
  • Professional solutions for employee safety
  • Easy to mount
  • Anti-limescale design and material
  • Improved functionality with long lifetime
  • Strong wall flange with 6 holes for wall mounted models with exposed piping
The water amount is regulated by a built-in FLOWFIX (14 L/min).
Min. working pressure: 1,5 bar.
Certifications: EN 15154 (Part 1); EN 15154 (Part 1, 2)