Imprimer
ASTM precision thermometers, H-B DURAC®, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
B607800200 B607800400
BELAB607800200EA 1800 EUR
BELAB607800200 BELAB607800400
ASTM precision thermometers, H-B DURAC®, SP Bel-Art
Thermomètres Thermomètres analogiques
These mercury free thermometers are individually calibrated at specified points.
  • Ideal for use in quality control laboratories
  • Blue organic liquid fill against lead-free white back glass, individually serialised
Calibration points: −7, 0, 10, 20, 30 °C (ASTM S63C-03); 0, 25, 35, 45, 55 °C (ASTM S64C-03).
Certifications: Conforms to ASTM standards for methods specified in the order table.
Informations de livraison: Supplied with a calibration report stating corrections to one-tenth of the smallest division.