Imprimer
DIN adapters for electrodes
Fournisseur: Mettler - Toledo
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
30403890 30499900
METT30403890EA 36.1 EUR
METT30403890 METT30499900
DIN adapters for electrodes
Câbles
These adapters will allow the LE electrodes with BNC to connect to meters with DIN.