Imprimer
Clear Polyester Component and Panel Labels, M211 and M210
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
110901 110899 110900 139748 139755 139749 139746 139747
BRDY110901EA 44.7 EUR
BRDY110901 BRDY110899 BRDY110900 BRDY139748 BRDY139755 BRDY139749 BRDY139746 BRDY139747
Clear Polyester Component and Panel Labels, M211 and M210
Étiquettes
Polyester labelling tape for label printers.
  • Translucent
  • High adhesion
  • Gloss finish
  • For smooth and textured/rough surfaces