Imprimer
Nylon Cloth Patch Panel and Wire and Cable Labels, M211 and M210
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
110938 110895 110894 110893
116-0627EA 30 EUR
116-0627 116-0617 116-0616 116-0615
Nylon Cloth Patch Panel and Wire and Cable Labels, M211 and M210
Étiquettes
Labelling tape made of nylon cloth.
  • Matt finish
  • For smooth, textured/rough or highly textured/low surface energy surfaces