Imprimer
Human pooled serum, French origin
Fournisseur: SEQENS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
21000P-500ML 21000P-100ML 21000P-1L
21000P-500MLEA 1970 EUR
21000P-500ML 21000P-100ML 21000P-1L
Human pooled serum, French origin
Sérums
This human pooled serum is obtained from off-the-clot serum units collected in France. Units are pooled and 0,2 μm filtered prior to being bottled. Possibility of gender and blood group selection.
  • Dry extract: 70 to 90 g/L
  • Total proteins: 60 to 80 g/L
  • Filtered through a 0,2 µm membrane
  • Shelf life: 3 years
Donations used for the manufacture of this product have been collected in EU licensed donation facilities, in accordance with ethical requirements of the French Code de la santé publique:
Donor consent: donations proceed from volunteer donors, who agreed their donation may be used in any way the collection centre deems appropriate
Gratuitousness: donations have not been paid
Anonymity: each donation is anonymous and is associated with a donation number
Each unit has also been tested and found negative or non-reactive for:
Anti-HIV 1+2 and anti-HCV antibodies
HBs antigen
HIV 1 and HCV RNA
Syphilitic serology
Complete donor traceability on file.
Attention: Only for research or manufacture of in vitro diagnostic reagents. Not for therapeutic use. This product of human origin should be considered as potentially infectious and handled accordingly.