Imprimer
ORP electrode solutions
Marques : SI Analytics
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
285138184 285138357 285138373 285138381 285138168
662-6171EA 82.6 EUR
662-6171 SCOI285138357 SCOI285138373 SCOI285138381 SCOI285138168
ORP electrode solutions
Étalons Étalons Redox/ORP
ORP electrode solutions are used as cleaning solutions for combination electrodes and reference electrodes.