Imprimer
Laboratory bottles, pressure resistant, glass
Fournisseur: NEUBERT VOLUME GLASSWAERE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0854-11-050004 0854-11-500045 0854-121000045 0854-16-5045
NEUB0854-11-050004EA 375 EUR
NEUB0854-11-050004 NEUB0854-11-500045 NEUB0854-121000045 NEUB0854-16-5045
Laboratory bottles, pressure resistant, glass
Flacons Flacons en verre
These bottles are made from pressure resistant borosilicate glass 3.3.
  • Without cap
  • Without spout
Pressure resistant from –1 to 1,5 bar
Certifications: ISO 4796.