Imprimer
General purpose thermometers, enclosed scale
Fournisseur: Amarell
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
G10016-FL G10000-FL G10000-FL-PTFE G10004-FL G10006-FL G10006-FL-PTFE G10008-FL G10010-FL G10010-FL-PTFE G10014-FL G10014-FL-PTFE
AMARG10016-FLEA 54.7 EUR
AMARG10016-FL AMARG10000-FL AMARG10000-FL-PTFE AMARG10004-FL AMARG10006-FL AMARG10006-FL-PTFE AMARG10008-FL AMARG10010-FL AMARG10010-FL-PTFE AMARG10014-FL AMARG10014-FL-PTFE
General purpose thermometers, enclosed scale
Thermomètres Thermomètres analogiques
These general purpose standard thermometers are designed for daily use in the laboratory. Capillary prismatic, colourless, total immersion.