Imprimer
Transfection reagent, in vivo SilenceMag™
Marques : OZ BIOSCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
IV-TK30240 IV-SM30500 IV-SM31000
OZBIIV-TK30240EA 154 EUR
OZBIIV-TK30240 OZBIIV-SM30500 OZBIIV-SM31000
Transfection reagent, in vivo SilenceMag™
Réactifs de transfection
In vivo SilenceMag™ is a rapid, simple and highly efficient method dedicated to transfection of small RNA (siRNA, miRNA) into target cells/tissue in vivo. It combines magnetic nanoparticles and small RNA that will be retained after injection at the magnetically targeted site. This targeted delivery method minimises systemic distribution, increases gene targeted inactivation and reduces toxicity. Furthermore, the magnetic forces enhance the uptake of magnetic nanoparticles by the target tissue, and thus improve the efficiency of silencing.
  • Increased silencing efficiency
  • Targeted process (magnetically-driven)
  • Reduction of the systemic dissemination of siRNA/miRNA during injection
  • Reduction of the siRNA/miRNA doses
  • Work under non permissive conditions (hypothermia, physiological flow conditions)
  • Penetration of the siRNA/miRNA into tissues
  • Minimum toxicity
Store at +4 °C.
Informations sur les accessoires: This reagent needs to be used with a magnetic plate.
Informations de livraison: Shipped at room temperature.