Imprimer
Stem transfection reagents, RmesFect™
Marques : OZ BIOSCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
RS30500 RS31000
OZBIRS30500EA 200 EUR
OZBIRS30500 OZBIRS31000
Stem transfection reagents, RmesFect™
Réactifs de transfection
RmesFect™ Stem transfection reagent is based on TEE-technology specifically designed for mRNA transfection in stem cells with high efficiency and low toxicity. It is specifically designed for in vitro mRNA (dsRNA) transfection in a large variety stem cells and primary cells.
  • Highly efficient, achieve mRNA delivery in all cells
  • Ready to use, no need for additional buffer
  • Low nucleic acid amount, minimised toxicity
  • Protects mRNA against degradation
  • Compatible with any culture medium
  • Serum compatible, medium change not required
mRNA transfection provides several advantages over plasmid DNA delivery, no need for nuclear uptake - mRNA translation into proteins occurs in the cytoplasm, faster protein expression than DNA transfection, no genomic integration, protein expression in a total promoter independent manner.
Store at –20 °C.
Informations de livraison: Shipped at room temperature.