Imprimer
DNA transfection reagent, PolyMag™
Marques : OZ BIOSCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PN30200 PN31000 PN30100
OZBIPN30200EA 241.76 EUR
OZBIPN30200 OZBIPN31000 OZBIPN30100
DNA transfection reagent, PolyMag™
Réactifs de transfection
PolyMag™ transfection reagent, based on Magnetofection™ transfection technology, has been optimised for use with all types of nucleic acids. The reagent is composed of magnetic nanoparticles coated with cationic molecules. It has been successfully tested on a large range of primary cells, hard to transfect cells and cell lines and its efficiency has been reported in many publications.
  • Highly efficient for primary cells
  • Powerful for hard to transfect cells
  • Non toxic
  • Serum-compatible
  • Simple, rapid and easy to use
  • For all types of nucleic acids
Store at +4°C.
Informations sur les accessoires: This reagent needs to be used with a magnetic plate.
Informations de livraison: Shipped at room temperature.