Imprimer
Quick-release clamps for flat flange connections
Fournisseur: Lenz Laborglas GmbH & CO.KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
06246010 06246015 06246020 06246006
201-0192EA 129 EUR
201-0192 LENZ06246015 201-0195 201-0191
Quick-release clamps for flat flange connections
Pinces Pinces à usage général
Stainless steel, rust-proof.
  • Simple assembly and disassembly
  • With three holding segments
  • Quick release
For clamping reaction vessels and lids together.