Imprimer
DNA transfection reagent, Verofect™
Marques : OZ BIOSCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VF60250 VF60500 VF61000
OZBIVF60250EA 137.6 EUR
OZBIVF60250 OZBIVF60500 OZBIVF61000
DNA transfection reagent, Verofect™
Réactifs de transfection
Verofect™ is a powerful transfection reagent designed to obtain highly efficient and reproducible transfection of Vero cells. It can be used for many applications such as stable and transient transfection, protein and viral production, etc.
  • Optimised for vero cells
  • Non toxic
  • Serum compatible
  • Simple, ready to use, and rapid
  • Highly efficient and reproducible results
Store at +4 °C.
Informations de livraison: Shipped at room temperature.