Imprimer
Transfection reagent, BrainFectIN
Marques : OZ BIOSCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
IV-BF30100 IV-BF30250 IV-BF30500
OZBIIV-BF30100EA 144.05 EUR
OZBIIV-BF30100 OZBIIV-BF30250 OZBIIV-BF30500
Transfection reagent, BrainFectIN
Transfection Reagents
BrainFectIN transfection reagent enables nucleic acid delivery into the central nervous system of small animals.
  • Targeted process (stereotaxic injection)
  • Good transfection efficiency
  • Reduction of the injection volume
  • Reduction of the DNA doses
  • Minimised toxicity
  • Low immunogenicity
  • Rapid and longterm transgene expression
Major difficulties with gene delivery into the central nervous system are a weakness of standard non viral gene carriers along with the limitations associated with use of viral particles (time-consuming and requires additional saftey precautions). Unlike these methods, BrainFectIN transfection reagent has an original non viral formulation that allows safe, easy and efficient nucleic acid delivery.
This transfection reagent allows transfection of neural cells in specific brain following stereotaxic injection, with low immunogenicity and rapid and longterm transgene expression.
Store at –20 °C.
Informations de livraison: Shipped at room temperature.