Imprimer
siRNA transfection reagent, SilenceMag™
Marques : OZ BIOSCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SM10200 SM10500 SM11000 SM13000 KC30300 KC30400
OZBISM10200EA 117.18 EUR
OZBISM10200 OZBISM10500 OZBISM11000 OZBISM13000 OZBIKC30300 OZBIKC30400
siRNA transfection reagent, SilenceMag™
Réactifs de transfection
SilenceMag™ siRNA delivery reagent is based on Magnetofection™ technology. Specifically designed for siRNA transfection, SilenceMag™ gives reliable high protein knockdown at very low doses of siRNA in numerous cell types (primary cells, hard-to-transfect and cell lines).
  • High protein knockdown efficiency
  • For all siRNA applications
  • No off-target effects
  • Serum compatible and non toxic
  • Simple, rapid, and easy to use
Store at +4 °C.
Informations sur les accessoires: Needs to be used with a magnetic plate, available as a separate accessory or as part of a starter kit.
Informations de livraison: Shipped at room temperature.