DNA transfection reagent, HYPE-293™
Marques : OZ BIOSCIENCESTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
HYPE-293™ transfection kits are dedicated to achieving high yield protein expression in HEK-293, 293-S, 293-F, or any 293-related cells grown in suspension.
- For optimal protein and antibody production yield
- Suitable for all HEK293 cells grown in suspension
- Compatible with any synthetic or regular media used for protein production
- Ideal for bioreactor, spinner, or flasks
- Animal-free origin
Scale up to larger volumes for production of milligrams of protein per litre of cell culture is straightforward, with simple and cost efficient handling steps.
Store HYPE-293 reagent and B293 reagent at –20 °C.
Informations de livraison: Shipped at room temperature.