Imprimer
DNA transfection reagent, HeLaFect™
Marques : OZ BIOSCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
HF20500 HF21000
OZBIHF20500EA 169.34 EUR
OZBIHF20500 OZBIHF21000
DNA transfection reagent, HeLaFect™
Réactifs de transfection
HeLaFect™ transfection reagent is specifically developed for HeLa cell lineage transfection with high efficiency. It is a lipid-based reagent based on Tee-Technology (Triggered Endosomal Escape). The cationic design of the reagent allows high nucleic acid compaction for efficient transport into HeLa cells. This reagent is composed of lipids, leading to high viability, and is ready to use.
  • Highly efficient, more than 80% of transfected HeLa cells
  • Ready-to-use, no need for additional buffer
  • Low nucleic acid amount minimised toxicity
  • High level of nucleic acid compaction
  • Easy and straight forward protocol
  • Compatible with any culture medium, medium change not required
Store at –20°C.
Informations de livraison: Shipped at room temperature.