Imprimer
J.T.Baker®, Female QT-2 Quick Connect Bodies
Fournisseur: Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
J.T.Baker®
9801-00EA 0 EUR
9801-00 9802-02 9802-03 9802-00 9802-01 9802-06 9802-07 9802-04 9802-05
J.T.Baker®, Female QT-2 Quick Connect Bodies
Raccords et connecteurs
Swagelock colour-coded quick connect for use with various solvents and gas ports.
  • For use with female tube fittings
  • For less than 1250 L sizes