Imprimer

Polyéthylèneglycol 6.000, en paillettes, Ultrapure

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Synonyme(s): PEG 6.000

J19972.A1 J19972.A7
J19972.A1EA 103 EUR
J19972.A1 J19972.A7
Polyéthylèneglycol 6.000, en paillettes, Ultrapure
Polyéthylèneglycol 6.000

Application includes differential precipitation of DNA, enhances hybridisation rate of nucleic acids, improves efficiency of end-labeling with T4 polynucleotide kinase, used in ligation of blunt ended DNA, precipitates bacteriophage ± particles and plasmid DNA, used in preparing single-stranded M13 DNA and promotes cell fusion.

Formule: H(OCH₂CH₂)nOH
Poids moléculaire: 6000 g/mol
Point de fusion: 60…63 °C
Température de stockage: Température ambiante
Numéro MDL: MFCD01779614
Numéro CAS: 25322-68-3
EINECS: 500-038-2
Indice Merck: 14,07568

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR