Imprimer
QuEChERS mixes, clean-up, CHROMABOND®
Fournisseur: MACHEREY-NAGEL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
730972 730973 730974 730975 730358.2 730646.2 730657 730657.2 730658 730669 730850.2 730858.2 730996.2 730670.2 730842 730843.2 730845
MANA730972EA 161 EUR
MANA730972 MANA730973 MANA730974 MANA730975 MANA730358.2 MANA730646.2 MANA730657 MANA730657.2 MANA730658 MANA730669 MANA730850.2 MANA730858.2 MANA730996.2 MANA730670.2 MANA730842 MANA730843.2 MANA730845
QuEChERS mixes, clean-up, CHROMABOND®
Colonnes pour chromatographie
CHROMABOND® QuEChERS mixes with special diamino silica phase are suitable for determination of pesticides in food samples. These remove polar compounds (organic acids, pigments, sugars) from matrices like fruit or vegetables.
  • Quick, Easy, Cheap, Effective, Rugged, Safe
  • Multi-residue pesticide analysis of fruits or vegetables
  • Pre-weighed CHROMABOND® QuEChERS mixes
For subsequent clean-up, a diamino phase (PSA) removes sugars and organic acids. Magnesium sulphate removes water, C18 ec removes nonpolar interferences such as fats and the carbon phase removes pigments, sterols, and nonpolar interferences.
For prior extraction see "QuEChERS mixes, extraction, CHROMABOND®".
CHROMABOND® QuEChERS mixes are weighed in 2 ml centrifuge tubes with snap cap or in 15 ml centrifuge tubes with screw cap.