Imprimer
Thermoreactors
Fournisseur: SELECTA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
4000638 4000639 4000640
SELE4000638EA 4820 EUR
SELE4000638 SELE4000639 SELE4000640
Thermoreactors
Chauffe-blocs Bains à sec numériques
Reflux method thermoreactor for the determination of Chemical Demand Oxygen "COD" in wastewater.
  • Uniform and simultaneous heating of samples
  • Automatic programming of temperature and digestion time
  • Acoustic and visual alarm at the end of the cycle
  • Alarm if the K type temperature probe fails.
Digestion block for 6, 12 or 20 samples, with process programmer of processes and lifting racks.
Racks with support tube holder in a chemically treated special plate, with handles and side plates to avoid heat losses.
For correct operation it is essential toEn savoir plus