Imprimer
ESD safety ankle boots, lace-up, Sander
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1768321/35 1768321/36 1768321/37 768321/38 768321/39 768321/40 768321/41 768321/42 768321/43 768321/44 768321/45 768321/46 768321/47
ELTN1768321/35EA 90.9 EUR
ELTN1768321/35 ELTN1768321/36 ELTN1768321/37 ELTN768321/38 ELTN768321/39 ELTN768321/40 ELTN768321/41 ELTN768321/42 ELTN768321/43 ELTN768321/44 ELTN768321/45 ELTN768321/46 ELTN768321/47
ESD safety ankle boots, lace-up, Sander
Chaussures et bottes
100% metal- and leather-free, lightweight ESD safety ankle boot made from hydrophobised microfibre and hydrophobised CORDURA® (tear and abrasion resistant, dirt and water repellent), a padded tongue plus a slip resistant, dual-density TPU/PU sole (Trainers).
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length fleece inlay sole (ESD Pro), black
  • Metal-free puncture resistant midsole
  • Composite toe protection cap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/red
Certifications: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC.