Imprimer
ESD safety ankle boots, lace-up, Nova, Impulse Lady Aqua Boa
Fournisseur: Elten
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Impulse Lady Aqua Boa®
ELTN74841/41EA 86.9 EUR
ELTN74841/41 ELTN74841/40 ELTN74841/42 ELTN74841/36 ELTN74841/35 ELTN74841/38 ELTN74841/37 ELTN74841/39
ESD safety ankle boots, lace-up, Nova, Impulse Lady Aqua Boa
Chaussures et bottes
Metal- and leather-free, lightweight ESD safety ankle boots made from perforated microfibre and reflective materials, skin-friendly textile lining, a full padded bellows tongue and collar and a dual-density PU/PU sole NOVA.
  • Climate regulating, breathable textile lining
  • Elastic lacing system with individual quick locking system
  • Replaceable insole; orthopaedic insoles possible
  • Full-length inlay sole LADIES ESD
  • Protective steel toecap (200 J)
  • Metal-free, puncture resistant midsole
Ideal for use in dry work areas such as industry, storage, transport, or assembly and where there is a risk of penetration from pointed and sharp objects. The shoe is tailored to the ergonomics of female feet.
Colour: Black/turquoise
Certifications: EN 61340 (ESD); DGUV 112-191; EN ISO 20345; S1P, SRC