Imprimer
ESD safety shoes, sandals, Nova, Impulse Lady Aqua Easy
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
74801/35 74801/36 74801/37 74801/38 74801/39 74801/40 74801/41 74801/42
ELTN74801/35EA 84.9 EUR
ELTN74801/35 ELTN74801/36 ELTN74801/37 ELTN74801/38 ELTN74801/39 ELTN74801/40 ELTN74801/41 ELTN74801/42
ESD safety shoes, sandals, Nova, Impulse Lady Aqua Easy
Chaussures et bottes
Metal- and leather-free, lightweight ESD safety sandals made from perforated microfibre and reflective materials, skin friendly textile lining, a full padded bellows tongue and collar plus a dual-density PU/PU sole (Nova).
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length inlay sole (Ladies ESD)
  • Protective steel toecap (200 J)
  • Metal-free, puncture resistant midsole
Ideal for use in dry work areas such as industry, storage, transport, or assembly and where there is a risk of penetration from pointed and sharp objects. Ergonomically tailored to female feet.
Colour: Black/turquoise
Certifications: EN 61340 (ESD); DGUV 112-191; EN ISO 20345; S1P, SRC