Imprimer
ESD safety shoes, Velcro® fastening, White Grip
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
729685/35 729685/36 729685/37 729685/38 729685/39 729685/40 729685/41 729685/42 729685/43 729685/44 729685/45 729685/46 729685/47 729685/48
ELTN729685/35EA 84.9 EUR
ELTN729685/35 ELTN729685/36 ELTN729685/37 ELTN729685/38 ELTN729685/39 ELTN729685/40 ELTN729685/41 ELTN729685/42 ELTN729685/43 ELTN729685/44 ELTN729685/45 ELTN729685/46 ELTN729685/47 ELTN729685/48
ESD safety shoes, Velcro® fastening, White Grip
Chaussures et bottes
Leather-free ESD safety shoe made from hydrophobised microfibre, a padded tongue and a dual-density rubber/PU sole (E-Track).
  • Climate regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length inlay (Sportive ESD), grey
  • Soft fleece ESD insole
  • Anatomically-shaped insole and ankle support for improved stability
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected.
Colour: White
Certifications: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S2, SRC