Imprimer
ESD safety shoes, lace-up, Malvin Blue
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
729541/40 729541/41 729541/42 729541/43 729541/44 729541/45 729541/46 729541/47 729541/48
ELTN729541/40EA 100.9 EUR
ELTN729541/40 ELTN729541/41 ELTN729541/42 ELTN729541/43 ELTN729541/44 ELTN729541/45 ELTN729541/46 ELTN729541/47 ELTN729541/48
ESD safety shoes, lace-up, Malvin Blue
Chaussures et bottes
ESD safety shoes made from hydrophobised nubuck leather with mesh inserts, a full padded bellows tongue plus a dual-density TPU/PU sole (Maxximo).
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length inlay sole (Sportive ESD)
  • Anatomically-shaped insole and ankle support for improved stability
  • Cushioning midsole made of Infinergy® by BASF rebounds back extremely well when under pressure giving the energy back to the wearer
  • Abrasion resistant scuff cap protects the leather upper
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/blue
Certifications: EN 61340-5-1 (ESD); EN ISO 20345; S3, SRC.