Imprimer
ESD safety shoes, lace-up, Safeguard, Senex Al
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
728551/38 728551/39 728551/40 728551/41 728551/42 728551/43 728551/44 728551/45 728551/46 728551/47 728551/48
ELTN728551/38EA 88.9 EUR
ELTN728551/38 ELTN728551/39 ELTN728551/40 ELTN728551/41 ELTN728551/42 ELTN728551/43 ELTN728551/44 ELTN728551/45 ELTN728551/46 ELTN728551/47 ELTN728551/48
ESD safety shoes, lace-up, Safeguard, Senex Al
Chaussures et bottes
Leather-free, sporty and lightweight ESD safety shoe made from hydrophobised microfibre and hydrophobised CORDURA® (tear and abrasion resistant, dirt and water repellent), a padded tongue plus a slip resistant, dual-density PU/PU tread sole Safeguard.
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Semi-orthopaedic inlay sole ESD
  • TPU scuff cap protects the leather upper
  • Metal-free, puncture resistant midsole
  • Aluminium toe cap (200 J)
  • Extra-wide version
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/red
Certifications: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC.