Imprimer
ESD safety shoes, lace-up, Maverick Black/Red
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
723391/48 723391/45 723391/46 723391/47 723391/42 723391/43 723391/44 723391/38 723391/39 723391/40 723391/41
ELTN723391/48EA 84.9 EUR
ELTN723391/48 ELTN723391/45 ELTN723391/46 ELTN723391/47 ELTN723391/42 ELTN723391/43 ELTN723391/44 ELTN723391/38 ELTN723391/39 ELTN723391/40 ELTN723391/41
ESD safety shoes, lace-up, Maverick Black/Red
Chaussures et bottes
Sporty, lightweight leather-free ESD safety shoes made from hydrophobised microfibre/hydrophobised textile material, a padded tongue plus a slip resistant, dual-density PU/PU sole (Fun).
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length inlay sole (ESD Pro), black
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/red
Certifications: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC.