Imprimer
ESD safety shoes, sandals, Velcro® fastening, Scott
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
72121/38 72121/39 72121/40 72121/41 72121/42 72121/43 72121/44 72121/45 72121/46 72121/47
ELTN72121/38EA 84.9 EUR
ELTN72121/38 ELTN72121/39 ELTN72121/40 ELTN72121/41 ELTN72121/42 ELTN72121/43 ELTN72121/44 ELTN72121/45 ELTN72121/46 ELTN72121/47
ESD safety shoes, sandals, Velcro® fastening, Scott
Chaussures et bottes
100% metal- and leather-free, lightweight ESD safety sandal made from microfibre and textile upper, reflective materials, a padded tongue and a slip-resistant dual-density TPU/PU sole SPORTICS.
  • Climate regulating, breathable textile lining
  • Full-length inlay sole ESD PRO black
  • Soft fleece ESD insole
  • Non-metallic puncture resistant midsole
  • Composite toe protection cap (200 J)
Ideal for use in dry work areas such as industry, storage, transport, or assembly and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certifications: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S1P, SRC