Imprimer
ESD safety shoes, sandals, Velcro® fastening, Scott
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
71872/35 71872/36 71872/37 71872/38 71872/40 71872/41 71872/42 71872/43 71872/44 71872/45 71872/46 71872/47 71872/48
ELTN71872/35EA 79.9 EUR
ELTN71872/35 ELTN71872/36 ELTN71872/37 ELTN71872/38 ELTN71872/40 ELTN71872/41 ELTN71872/42 ELTN71872/43 ELTN71872/44 ELTN71872/45 ELTN71872/46 ELTN71872/47 ELTN71872/48
ESD safety shoes, sandals, Velcro® fastening, Scott
Chaussures et bottes
100% metal- and leather-free, lightweight ESD safety sandal made from microfibre and textile upper, reflective materials, a padded tongue plus a slip resistant, dual-density TPU/PU sole (Sportics).
  • Climate-regulating, breathable textile lining
  • Full length inlay sole (ESD Pro), black
  • Soft fleece ESD insole
  • Composite toe protection cap (200 J)
Ideal for use in dry work areas such as industry, storage, transport, or assembly.
Colour: Black
Certifications: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S1, SRC.