Imprimer
ESD safety ankle boots, lace-up, Biomex Dynamics, Impulse Green
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
762551/36 762551/37 762551/38 762551/39 762551/40 762551/41 762551/42 762551/43 762551/44 762551/45 762551/46 762551/47
ELTN762551/36EA 92.9 EUR
ELTN762551/36 ELTN762551/37 ELTN762551/38 ELTN762551/39 ELTN762551/40 ELTN762551/41 ELTN762551/42 ELTN762551/43 ELTN762551/44 ELTN762551/45 ELTN762551/46 ELTN762551/47
ESD safety ankle boots, lace-up, Biomex Dynamics, Impulse Green
Chaussures et bottes
Metal- and leather-free, lightweight ESD safety ankle boots made from perforated microfibre and reflective materials, skin-friendly textile lining, a full padded bellows tongue and collar and an extremely flexible, ergonomically S-shaped dual-density TPU/PU sole BIOMEX DYNAMICS.
  • Climate regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Semi-orthopaedic, anatomically shaped insole supports the natural heel-to-toe movement of the foot
  • Full-length ESD inlay sole
  • Abrasion-proof scuff cap
  • Protective composite toecap (200 J)
  • Metal-free, puncture resistant midsole
Ideal for use in dry work areas such as industry, storage, transport, or assembly and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/green
Certifications: EN 61340 (ESD); DGUV 112-191; EN ISO 20345; S1P, SRC