Imprimer
Anti-CN37 Mouse Monoclonal Antibody [clone: 1H10]
Fournisseur: Biosensis
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BSENM-1802-50EA 302 EUR
BSENM-1802-50
Anti-CN37 Mouse Monoclonal Antibody [clone: 1H10]
Anticorps
May participate in RNA metabolism in the myelinating cell, CNP is the third most abundant protein in central nervous system myelin. Ref: uniprot.org
Western blotting (1:5,000-1:20,000) and Immunocytochemistry (1:1,000-1:5,000). Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: CN37
Clonality: Monoclonal
Clone: 1H10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat