Imprimer
IV cannulae, Vasofix® Certo
Fournisseur: B. Braun
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
4269098 4269110
720-2550EA 149 EUR
720-2550 720-2551
IV cannulae, Vasofix® Certo
Cannulae
Vasofix Certo IV catheter with injection port. IV catheter material available in PU and FEP for low-pain, gentle insertion of catheter. Integrated injection port: Medication can be quickly administered without having to repuncture the area.
  • Four fully integrated x-ray contrast lines
  • Hydrophobic blood stoppers, removable lock cones
  • Colour-coded sizes
  • Free of latex and PVC
  • Excellent puncture properties with atraumatic tip and triple facet bevel geometry
Same product properties as the Vasofix Safety (with FEP catheter) excluding the integrated, self-activating safety device, for preventing needlestick injuries and the associated risk of infection.
Certifications: ISO 10555-1/5.