Imprimer
Tablet counting triangles, SteriWare®
Fournisseur: Sampling Systems
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8066A-1 8066A-1S
SAMP8066A-1EA 231 EUR
SAMP8066A-1 SAMP8066A-1S
Tablet counting triangles, SteriWare®
Plateaux/Récipients/Pots Plateaux et récipients de laboratoire
The SteriWare® counting triangles are ideal for counting round tablets. They are manufactured and packed in a cleanroom and so can be used straight from the bag.
  • Made from FDA conforming PS
  • BSE/TSE free
  • Suitable for left handed and right handed operators
  • With an embossed scale to aid operation
Certifications: Conforms to FDA, EU 10/2011 and EC 1935/2004. Sterility Assurance Level (SAL) of 10⁻⁶.
Cdt: Individually bagged in a heat-sealed laminate bag.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable