Imprimer
Safety ankle boots, lace-up, Basic Compo Mid, Jori
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
16111/36 16111/37 16111/38 16111/39 16111/40 16111/41 16111/42 16111/43 16111/44 16111/45 16111/46 16111/47 16111/48
ELTN16111/36EA 43.9 EUR
ELTN16111/36 ELTN16111/37 ELTN16111/38 ELTN16111/39 ELTN16111/40 ELTN16111/41 ELTN16111/42 ELTN16111/43 ELTN16111/44 ELTN16111/45 ELTN16111/46 ELTN16111/47 ELTN16111/48
Safety ankle boots, lace-up, Basic Compo Mid, Jori
Chaussures et bottes
100% metal-free, extremely lightweight safety ankle boot made from cowhide leather, reflective materials, skin-friendly textile lining, a full padded bellows tongue and collar padding and a dual-density PU/PU jo_BASIC sole (oil, fuel and abrasion resistant, heat resistant up to approx. 130 °C).
  • Climate regulating, breathable textile lining
  • Full-length inlay sole
  • PU toe protection cap
  • Puncture resistant composite midsole
  • Protective composite toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects as well as areas close to induction loops or metal detectors.
Colour: Black
Certifications: EN ISO 20345; S3, SRC