Imprimer
Safety ankle boots, BOA® Fit System, jo_Max BOA® Mid, Jori
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
16771/38 16771/39 16771/40 16771/41 16771/42 16771/43 16771/44 16771/45 16771/46 16771/47 16771/48 16771/36 16771/37
ELTN16771/38EA 88.9 EUR
ELTN16771/38 ELTN16771/39 ELTN16771/40 ELTN16771/41 ELTN16771/42 ELTN16771/43 ELTN16771/44 ELTN16771/45 ELTN16771/46 ELTN16771/47 ELTN16771/48 ELTN16771/36 ELTN16771/37
Safety ankle boots, BOA® Fit System, jo_Max BOA® Mid, Jori
Chaussures et bottes
Anti-static safety ankle boot made from hydrophobised nubuck leather, mesh inserts and reflective materials, skin friendly textile lining, a full, padded bellows tongue plus collar, and a dual-density rubber/PU jo_Max sole (3,0 mm profile, oil, fuel and chemical resistant, also temperature resistant from approx. –20 to +200 °C).
  • Climate-regulating, breathable textile lining
  • Heel pocket lining made from sturdy abrasion resistant microfibre lining
  • Full length inlay sole
  • Lace- and buckle-free Boa® system can be opened and closed with just one hand
  • Protective composite toecap (200 J)
  • Metal-free, puncture resistant midsole
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certifications: EN ISO 20345; S3, SRC