Imprimer
Volumetric flasks, class A, trapezoidal shape, DAkkS, BLAUBRAND®
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
DAKKS36401 DAKKS36402
BRNDDAKKS36401EA 121 EUR
BRNDDAKKS36401 BRNDDAKKS36402
Volumetric flasks, class A, trapezoidal shape, DAkkS, BLAUBRAND®
Ballons Fioles jaugées
Borosilicate glass 3.3, with PP stopper.
  • Calibrated to contain (TC, In)
  • Marks and inscriptions in high contrast blue enamel
  • DE-M marking
*Wide neck
Certifications: DIN EN ISO 1042
Informations de commande: Supplied with DAkkS calibration certificate.