Imprimer
Low noise data acquisition system plus HumSilencer, Digidata®
Fournisseur: Molecular Devices
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
DIGIDATA1550B0 DIGIDATA1550B1 DIGIDATA1550B4
MLDVDIGIDATA1550B0EA 0 EUR
MLDVDIGIDATA1550B0 MLDVDIGIDATA1550B1 MLDVDIGIDATA1550B4
Low noise data acquisition system plus HumSilencer, Digidata®
Sonomètres
The low noise data acquisition system plus HumSilencer™ adaptive noise cancellation with the added benefit of single-click elimination of 50/60 Hz line-frequency noise up to four channels.
  • Built-in, softwarecontrolled technology
  • Removes local line-frequency noise patterns
  • Associated high-frequency harmonics from incoming signals in less than one second
Intended for precision scientific applications, it is particularly designed for electrophysiology experiments, to send and receive signals from microelectrode amplifiers, and to interact with peripheral instruments such as solution exchangers.