Imprimer
Swinging agitator
Fournisseur: TOULEMONDE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AG6A AG5 AG3 AG6B
445-0006EA 7430 EUR
445-0006 445-0005 TOULAG3 TOULAG6B
Swinging agitator
Agitateurs Agitateurs orbitaux
Designed to solve the problem of agitation of separating funnels in laboratories operating liquid phase separations.
  • Continuous speed variator, allowing to choose the appropriate speed
  • Eliminates foaming, emulsion or destruction of macro-molecules
  • Timer for setting stirring times up to 30 minutes
The tray of this device has a vertical position for gravity settling and easy removal of separate components.