Imprimer
Titrators, SIMPLEX
Fournisseur: GERBER FUNKE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
4520 4521
GERB4520EA 545 EUR
GERB4520 GERB4521
Titrators, SIMPLEX
Titrateurs
These SIMPLEX titrators consist of a polyethylene bottle on a plastic base.
  • Burette with automatic zero point adjustment
  • Precision titration by button control
Used for milk and cream.
Informations de livraison: Supplied with one 1 ml and one 25 ml pipette, one 200 ml Erlenmeyer flask.