Imprimer
Calcium ionophore II for ion selective electrodes, Selectophore™, Supelco®
Marques : Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
21193-250MG-F 21193-50MG-F
21193-250MG-FEA 824 EUR
21193-250MG-F 21193-50MG-F
Calcium ionophore II for ion selective electrodes, Selectophore™, Supelco®
Électrolytes Solutions de remplissage d'électrode
Lipophilic ionophore for Ca²⁺ selective electrodes with detection limit in the sub-nanomolar range.
Extremely efficient carrier of Ca²⁺ -ions; induces a selectivity in membranes for Ca²⁺ over Mg²⁺ by a factor of about 10⁴. Calcium ionophore II (ETH 129) was used in preparing calcium selective minielectrodes which are used in determination of dissociation constant (Kd).
Cdt: Packed in bottomless glass bottle. Contents are inside inserted fused cone.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79dce6fa0178eb2f82e1641265ee048bc95a06bb&vid=79dce6fa0178eb2f82e1641265ee048bc95a06bb&prid=web&clid=SAPDAM