Imprimer

6-Chloropurine 99+%

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Attention

Synonyme(s): 6-Chloro-7H-purine, 6-Chloro-1H-purine, 6-Chloro-9H-purine

171280050 171280250
ACRO171280050EA 137 EUR
ACRO171280050 ACRO171280250
6-Chloropurine 99+%
6-Chloropurine
Formule: C₅H₃ClN₄
Point d'ébullition: 185 - 192 °C (2 -rr)
Point de fusion: >300 °C (déc.)
Température de stockage: Température ambiante
Numéro MDL: MFCD00075825
Numéro CAS: 87-42-3
EINECS: 201-745-6
Indice Merck: 13,02180

Commander maintenant

Résultats des tests de spécification

Appearance Yellow to orange Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (105°C, 5 hrs)
Sulfated ash ≤0.1 % (800°C)
Appearance of solution (0.1% soln. in 0.1 M HCl) clear, opalescent to yellowish
Titration with HClO4 99.0 to 101.0 %
Carbon (C) 38.47 to 39.25 % (On dry substance)
Nitrogen (N) 32.52 to 36.97 % (On dry substance)
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR