Imprimer
Cut protection gloves, uvex C500 dry
Fournisseur: UVEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6049907 6049908 6049909 6049910 6049911
UVEX6049907EA 205 EUR
UVEX6049907 UVEX6049908 UVEX6049909 UVEX6049910 UVEX6049911
Cut protection gloves, uvex C500 dry
Gants
The uvex C500 series sets standards regarding protection, comfort, flexibility, tactility and economy. This high-tech safety glove combines all of these qualities with the highest level of cut protection, level 5/C.
  • Highly flexible
  • Palm and fingers with high-performance vinyl (HPV) grip
  • Excellent grip in dry environments
  • Very good tactile sensation
  • Very high level of cut protection with patented uvex Bamboo TwinFlex® technology
  • Silicone-free according to imprint test
  • Highly breathable thanks to uvex climazone
Ideal for use in assembly, maintenance, metalworking, shipping/logistics, sorting, glass handling, metal parts processing and the automotive, paper, iron/steel, building and construction industries.
Certifications: EN 388:2016 (XX4XC); Öko-Tex Standard 100