Imprimer
Cleaning agents for use in ultrasonic baths, TICKOPUR TR3, Bandelin/basan
Fournisseur: BANDELIN-ELECTRONIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
923 913 935
142-2015EA 49.3 EUR
142-2015 BAND913 BAND935
Cleaning agents for use in ultrasonic baths, TICKOPUR TR3, Bandelin/basan
Produits nettoyants Détergents et Nettoyants de Surfaces
The cavitation improving and material protecting features allow a maximum effect of the ultrasonic cleaning.
  • Shortens the cleaning time significantly
  • Special cleaner based on citric acid, mildly acidic
  • Free from phosphate and have corrosion protection
Used for cleaning, rust removal, decalcification and degreasing of assembled and disassembled machines, tools and devices made of steel, stainless steel, non-ferrous, precious and light metals, glass, ceramics, plastics and rubber. Contaminants such as mineral residues, drifting rust, grease, oils, waxes, pigments, drilling, grinding, polishing and lapping residues can be cleaned.