Imprimer
Cuivre (II) oxyde ≥97%
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Attention
Synonyme(s): Oxyde de cuivre (II)
12299.36 12299.A3 12299.A9
12299.36EA 101 EUR
12299.36 12299.A3 12299.A9
Cuivre (II) oxyde ≥97%
Cuivre (II) oxyde
Purity: 97% CAS No.: 1317-38-0 Molecular Formula: CuO Formula Weight: 79.55 Physical Form: -325 Mesh Powder Density: 6.3 Refractive Index: 2.63 Appearance: Black Odour: Odourless Melting Point: 1326° Solubility: Insoluble in water and alcohol. Soluble in dil. acids, NH4Cl and KCN Merck Reference: 14,2646 UN No.: 3077 MDL No.: MFCD00010979
As a pigment, in sweeting petroleum gases, in galvanic electrodes, metallurgical flux, as an optical glass polishing agent, and as a catalyst in organic reactions
Formule: CuO
Poids moléculaire: 79.55 g/mol
Point d'ébullition: 2000
Point de fusion: 1326
Densité: 6.3
Température de stockage: Température ambiante
Numéro MDL: MFCD00010979
Numéro CAS: 1317-38-0
EINECS: 215-269-1
UN: 3077
ADR: 9 III
Indice Merck: 13,02674