Imprimer
Neodymium(III) chloride hydrate ≥99.9% (REO, base d'oxyde de terre rare), REacton®
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Attention
Synonyme(s): Neodymium trichloride hydrate
11251.18 11251.30
11251.18EA 143 EUR
11251.18 11251.30
Neodymium(III) chloride hydrate ≥99.9% (REO, base d'oxyde de terre rare), REacton®
Neodymium(III) chloride hydrate
Neodymium(III) chloride hydrate is used for production of neodymium metal. It also finds application in lasers and fibre amplifiers. It serves as dopants for silica based as well as plastic fibres. It plays an important role of additives in organic light emitting diodes resulting in improved LED emission spectrum. In addition, it is used in catalysis of polymerisation of various dienes. It is used to modify titanium dioxide for use as a photocatalyst. Its coating on aluminium and its alloy protect them from corrosion. It is also extensively used as fluorescent labels for organic molecules.
Purity: 99.9% CAS No.: 10024-93-8 Molecular Formula: NdCl3.xH2O Formula Weight: 250.60(anhy) Physical Form: Crystalline Aggregates Odour: Odourless Merck Reference: 14,6450 MDL No.: MFCD00011132
Formule: NdCl₃·xH₂O
Poids moléculaire: 250.60 g/mol (anhy)
Point de fusion: 124 °C
Densité: 2.28 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00011132
Numéro CAS: 13477-89-9
EINECS: 233-031-5
Indice Merck: 14,6450